Potassium chloride, 0.0765M, Conductivity Standard
Potassium chloride, 0.0765M, Conductivity Standard
Potassium chloride, 0.0765M, Conductivity Standard
Thermo Scientific Chemicals

Potassium chloride, 0.0765M, Conductivity Standard

CAS: 7447-40-7 | ClK | 74.55 g/mol
数量:
500 mL
1 L
4 L
製品番号(カタログ番号) 042695.K2
または、製品番号042695-K2
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
仕様
CAS7447-40-7
ChEBICHEBI:32588
化学物質名または材質Potassium chloride Conductivity Standard
EINECS番号231-211-8
式量74.55
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only