Polystyrene latex microsphere, 15.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 15.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 15.0 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 15.0 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
製品番号(カタログ番号)数量
042718.AA
または、製品番号042718-AA
5 mL
製品番号(カタログ番号) 042718.AA
または、製品番号042718-AA
価格(JPY)
-
数量:
5 mL
一括またはカスタム形式をリクエストする
仕様
CAS9003-53-6
ChEBICHEBI:27452
化学物質名または材質Polystyrene latex microsphere
Concentration or Composition (by Analyte or Components)2.5 wt% dispersion in water
IUPAC名styrene
さらに表示
Polystyrene latex microsphere is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery. And also used in protein binding, size markers. The most popular use of latex beads is in the diagnostic use of adsorbed antibodies or antigens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only