Nickel Chromium wire, 1.6mm (0.064 in.) dia., Thermo Scientific Chemicals
Nickel Chromium wire, 1.6mm (0.064 in.) dia., Thermo Scientific Chemicals
Nickel Chromium wire, 1.6mm (0.064 in.) dia., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel Chromium wire, 1.6mm (0.064 in.) dia., Thermo Scientific Chemicals

製品番号(カタログ番号)数量
042838.G6
または、製品番号042838-G6
10 m
製品番号(カタログ番号) 042838.G6
または、製品番号042838-G6
価格(JPY)
-
数量:
10 m
一括またはカスタム形式をリクエストする
化学物質識別子
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
さらに表示
Comment1.6 mm diameter
CommentNi 77% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only