Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles
Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles
Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles

CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol
数量:
100 mL
500 mL
製品番号(カタログ番号) 042859.AE
または、製品番号042859-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100-58-3
IUPAC Namebromo(phenyl)magnesium
Molecular FormulaC6H5BrMg
InChI KeyANRQGKOBLBYXFM-UHFFFAOYSA-M
SMILESBr[Mg]C1=CC=CC=C1
さらに表示
FormClear liquid
Assay (Titration)2.7 to 3.3 M
Phenylmagnesium bromide is an important reagent called as Grignard reagent used in the synthesis of organic and oraganometallic reactions. It is used to prepare phenyl aryl sulphides, sulphoxides, and sulphones. It acts as a strong acid and Lewis acid and undergoes addition reaction with many unsaturated functional groups. It is involved in the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s, 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, o-substituted benzophenones and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol. In addition to this, it is used for the monalkylation of primary amines to get secondary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only