Chromium granules, -4+7 mesh, 99.999% (metals basis)
Chromium granules, -4+7 mesh, 99.999% (metals basis)
Chromium granules, -4+7 mesh, 99.999% (metals basis)
Thermo Scientific Chemicals

Chromium granules, -4+7 mesh, 99.999% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
製品番号(カタログ番号) 042926.18
または、製品番号042926-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
さらに表示
FormGranules,
Particle Size-4+7 mesh
Assay from Supplier's CofA≥99.998% metals basis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only