Thermo Scientific Chemicals

Titanium(II) oxide pellets, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 042940.22
100 g, Each
Thermo Scientific Chemicals

Titanium(II) oxide pellets, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 042940.22
100 g, Each
Quantity
Catalog number: 042940.22
also known as 042940-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
12137-20-1
IUPAC Name
oxotitanium
Molecular Formula
OTi
InChI Key
OGIDPMRJRNCKJF-UHFFFAOYSA-N
SMILES
O=[Ti]
Total Metal Impurities
0.1% max.

Description

Used for vacuum deposition. Also applications include sunscreens and UV-blocking in cosmetics, varnishes, wood preservatives, textile fibers, and packaging films. Also used as paint pigment, opacifying agent, welding rod fluxes, optical coatings, as a catalyst, ceramic finish coat, plastics, elastomers, coated fabrics, printing inks, roofing granules, glass, and in glazes. Also identified as Titanium monoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text