Ruthenium(III) nitrosylchloride hydrate
Ruthenium(III) nitrosylchloride hydrate
Ruthenium(III) nitrosylchloride hydrate
Ruthenium(III) nitrosylchloride hydrate
Thermo Scientific Chemicals

Ruthenium(III) nitrosylchloride hydrate

CAS: 32535-76-5 | Cl3NORu | 237.42 g/mol
製品番号(カタログ番号) 043089.06
または、製品番号043089-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS32535-76-5
IUPAC Nametrichloro(nitroso)rutheniumtris(ylium)
Molecular FormulaCl3NORu
InChI KeyXTHALOZYNGMAKY-UHFFFAOYSA-K
SMILESCl[Ru+3](Cl)(Cl)N=O
さらに表示
Assay (unspecified)Ru 39% min
Used as catalyst. As a precursor to other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only