Lanthanum(III) sulfate hydrate, 99.9% (REO)
Lanthanum(III) sulfate hydrate, 99.9% (REO)
Lanthanum(III) sulfate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum(III) sulfate hydrate, 99.9% (REO)

CAS: 57804-25-8 | La2O12S3 | 565.98 g/mol
製品番号(カタログ番号) 043106.36
または、製品番号043106-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS57804-25-8
IUPAC Namedilanthanum(3+) trisulfate
Molecular FormulaLa2O12S3
InChI KeyVQEHIYWBGOJJDM-UHFFFAOYSA-H
SMILES[La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
Used as colorant or laboratory reagent. It is also mainly applied in specialty glass, phosphors, water treatment and catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only