Magnesium oxide, 2 part ceramic adhesive paste
Magnesium oxide, 2 part ceramic adhesive paste
Magnesium oxide, 2 part ceramic adhesive paste
Thermo Scientific Chemicals

Magnesium oxide, 2 part ceramic adhesive paste

High Temperature Adhesive For Bonding Ceramic To Metal
製品番号(カタログ番号) 043135.A4
または、製品番号043135-A4
価格(JPY)
-
数量:
2.5 kg
一括またはカスタム形式をリクエストする
仕様
CAS1309-48-4
化学物質名または材質Magnesium oxide
健康被害1H314-H335-H303: Causes severe skin burns and eye damage. May cause respiratory irritation. May be harmful if swallowed.
IUPAC名oxomagnesium
InChIキーCPLXHLVBOLITMK-UHFFFAOYSA-N
さらに表示
High temperature adhesive for bonding ceramic to metal

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C. Components: Magnesium oxide, 2 part ceramic adhesive paste (L) and Magnesium oxide, 2 part ceramic adhesive paste (P)
RUO – Research Use Only