Thermo Scientific Chemicals

Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals

Catalog number: 043182.14
25 g, Each
Thermo Scientific Chemicals

Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals

Catalog number: 043182.14
25 g, Each
Quantity
Catalog number: 043182.14
also known as 043182-14
Price (USD)
Price: 213.00
Online price: 181.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7789-02-8
IUPAC Name
chromium(3+) trinitrate
Molecular Formula
CrN3O9
InChI Key
PHFQLYPOURZARY-UHFFFAOYSA-N
SMILES
[Cr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (special)
?2.5 % of Theoretical value (Cr)
Form
Crystals
Total Metal Impurities
≤0.01% (100 ppm)
Appearance (Color)
Violet

Description

Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text