Thermo Scientific Chemicals

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 043204.30
250 g, Each
Thermo Scientific Chemicals

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 043204.30
250 g, Each
Quantity
Catalog number: 043204.30
also known as 043204-30
Price (USD)
Price: 598.00
Online price: 508.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2495-35-4
IUPAC Name
benzyl prop-2-enoate
Molecular Formula
C10H10O2
InChI Key
GCTPMLUUWLLESL-UHFFFAOYSA-N
SMILES
C=CC(=O)OCC1=CC=CC=C1
Assay (GC)
> 96.5%
Form
liquid
Appearance (Color)
Clear colorless to pale yellow

Description

Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text