Tin foil, 0.5mm (0.02in) thick, 99.8% (metals basis)
Tin foil, 0.5mm (0.02in) thick, 99.8% (metals basis)
Tin foil, 0.5mm (0.02in) thick, 99.8% (metals basis)
Thermo Scientific Chemicals

Tin foil, 0.5mm (0.02in) thick, 99.8% (metals basis)

Form: Approx. 9.1g/50 x 50mm | CAS: 7440-31-5 | Sn | 118.71 g/mol
数量:
50 x 50 mm
100 x 100 mm
100 x 500 mm
製品番号(カタログ番号) 043234.HP
または、製品番号043234-HP
価格(JPY)
-
数量:
100 x 500 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
さらに表示
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.8% (metals basis)
FormFoil, 0.5mm (0.02in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only