Thermo Scientific Chemicals

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 043261.KW
50 Ea., Each
Thermo Scientific Chemicals

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 043261.KW
50 Ea., Each
Quantity
Catalog number: 043261.KW
also known as 043261-KW
Price (USD)

Chemical Identifiers

CAS
124-40-3
IUPAC Name
dimethylamine
Molecular Formula
C2H7N
InChI Key
ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES
CNC
Form
Liquid
Assay from Supplier's CofA
39.0-41.0%
Appearance (Color)
Clear colorless

Description

Dimethylamine is a used as a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrochemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text