Thermo Scientific Chemicals

Silver(I) oxide, Electrical Grade, Thermo Scientific Chemicals

Catalog number: 043268.A1
1 kg, Each
Thermo Scientific Chemicals

Silver(I) oxide, Electrical Grade, Thermo Scientific Chemicals

Catalog number: 043268.A1
1 kg, Each
Quantity
Catalog number: 043268.A1
also known as 043268-A1
Price (USD)
Price: 3,713.00
Online price: 3,152.65
Your price:
Quantity
-

Chemical Identifiers

CAS
20667-12-3
IUPAC Name
(argentiooxy)silver
Molecular Formula
Ag2O
InChI Key
KHJDQHIZCZTCAE-UHFFFAOYSA-N
SMILES
[Ag]O[Ag]
Assay (unspecified)
Ag 90% min

Description

For battery plates, and pollution control filters. It is used in the preparation of other silver compounds, and silver-oxide batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text