2-Butanol, anhydrous, 99%
2-Butanol, anhydrous, 99%
2-Butanol, anhydrous, 99%
Thermo Scientific Chemicals

2-Butanol, anhydrous, 99%

Hygroscopic; H2O <0.01% | CAS: 78-92-2 | C4H10O | 74.12 g/mol
製品番号(カタログ番号) 043315.AK
または、製品番号043315-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
外見Colorless liquid
定量パーセント範囲99%
バイルシュタイン1718765
沸点98°C to 101°C
CAS78-92-2
さらに表示
2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only