L-Aspartic acid, 99%, low metals content
L-Aspartic acid, 99%, low metals content
L-Aspartic acid, 99%, low metals content
L-Aspartic acid, 99%, low metals content
Thermo Scientific Chemicals

L-Aspartic acid, 99%, low metals content

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol
製品番号(カタログ番号) 043317.36
または、製品番号043317-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS56-84-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
さらに表示
Assay (unspecified)> 98.5%
FormPowder
Loss on Drying< 0.2 %
Appearance (Color)White
L-Aspartic acid is used as a component of parenteral and enteral nutrition and as a pharmaceutical ingredient. it is used for cell culture and in manufacturing processes. It is widely utilized for mineral supplementation in the salt form.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only