Methyl levulinate, 99+%
Methyl levulinate, 99+%
Methyl levulinate, 99+%
Thermo Scientific Chemicals

Methyl levulinate, 99+%

CAS: 624-45-3 | C6H10O3 | 130.143 g/mol
製品番号(カタログ番号) 043397.14
または、製品番号043397-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS624-45-3
IUPAC Namemethyl 4-oxopentanoate
Molecular FormulaC6H10O3
InChI KeyUAGJVSRUFNSIHR-UHFFFAOYSA-N
SMILESCOC(=O)CCC(C)=O
さらに表示
Appearance (Color)Colorless
FormLiquid
Assay from Supplier's CofA≥99.0%
Methyl levulinate is mainly used as raw material on perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only