Thermo Scientific Chemicals

Cadmium iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043442.22
100 g, Each
Thermo Scientific Chemicals

Cadmium iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043442.22
100 g, Each
Quantity
Catalog number: 043442.22
also known as 043442-22
Price (USD)

Chemical Identifiers

CAS
7790-80-9
IUPAC Name
cadmium(2+) diiodide
Molecular Formula
CdI2
InChI Key
OKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES
[Cd++].[I-].[I-]
Total Metal Impurities
0.001% max.

Description

Cadmium iodide is used in lithography, photography, electroplating and the manufacturing of phosphors, in engraving process, and was used as an antiseptic and astringent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text