Titanium foil, 0.5mm (0.02in) thick, annealed, 99% (metals basis)
Titanium foil, 0.5mm (0.02in) thick, annealed, 99% (metals basis)
Titanium foil, 0.5mm (0.02in) thick, annealed, 99% (metals basis)
Thermo Scientific Chemicals

Titanium foil, 0.5mm (0.02in) thick, annealed, 99% (metals basis)

Form: approx. 22.5g/100x100mm | CAS: 7440-32-6 | Ti | 47.87 g/mol
数量:
100 x 100 mm
200 x 200 mm
200 x 1000 mm
製品番号(カタログ番号) 043677.GW
または、製品番号043677-GW
価格(JPY)
-
数量:
200 x 200 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
さらに表示
Appearance (Color)Silvery.
FormFoil, 0.5mm (0.02in) thick, annealed.
Assay from Supplier's CofA≥99.0% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only