Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
Thermo Scientific Chemicals

Chloroform, HPLC Grade, 99.5+% min, stab. with amylene

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
数量:
1 L
4 L
4 x 1 L
4 x 4 L
製品番号(カタログ番号) 043685.K7
または、製品番号043685-K7
価格(JPY)
-
数量:
4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
さらに表示
Optical Absorbance at245nm: 1.00AU, max
CommentSubmicron filtered
Impurity contentAcetone and aldehyde ( as acetone): Passes Test
Optical Absorbance at270nm: 0.02AU, max
Optical Absorbance at290nm: 0.01AU, max
さらに表示
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only