Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 99+%

Catalog number: 043701.22
100 g, Each
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 99+%

Catalog number: 043701.22
100 g, Each
Quantity
Catalog number: 043701.22
also known as 043701-22
Price (USD)
Price: 104.00
Online price: 88.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-26-9
IUPAC Name
N-[(prop-2-enamido)methyl]prop-2-enamide
Molecular Formula
C7H10N2O2
InChI Key
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES
C=CC(=O)NCNC(=O)C=C
Assay (unspecified)
> 99.0%

Description

Bisacrylamide is used in biochemistry as a preparatory agent for polyacrylamide gel for electrophoretic separation of nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text