Thermo Scientific Chemicals

Cloruro de niobio(V), Puratronic™, 99,999 % (base metálica), Thermo Scientific Chemicals

Número de catálogo: 043729.22
100 g, Each
Thermo Scientific Chemicals

Cloruro de niobio(V), Puratronic™, 99,999 % (base metálica), Thermo Scientific Chemicals

Número de catálogo: 043729.22
100 g, Each
Cantidad
Número de catálogo: 043729.22
también denominado 043729-22
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
10026-12-7
IUPAC Name
niobium(5+) pentachloride
Molecular Formula
Cl5Nb
InChI Key
YHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
Total Metal Impurities
0.001% max.

Descripción

Niobium(V) chloride is the precursor in the preparation of pure niobium, and ferroniobium. It is used as a Lewis acid catalyst in organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Envasado bajo argón
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H314 - Causes severe skin burns and eye damage

    EU Specific Hazard Statements

    EUH014 - Reacts violently with water

    EUH029 - Contact with water liberates toxic gas

    Precautionary Statements

    P402 + P404 - Store in a dry place. Store in a closed container

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician