Thermo Scientific Chemicals

Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043731.22
100 g, Each
Thermo Scientific Chemicals

Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043731.22
100 g, Each
Quantity
Catalog number: 043731.22
also known as 043731-22
Price (USD)
Price: 835.00
Online price: 709.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7721-01-9
IUPAC Name
tantalum(5+) pentachloride
Molecular Formula
Cl5Ta
InChI Key
OEIMLTQPLAGXMX-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
Total Metal Impurities
0.005% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text