Titanium gauze, 18 mesh woven from 0.28mm (0.011in) dia wire
Titanium gauze, 18 mesh woven from 0.28mm (0.011in) dia wire
Titanium gauze, 18 mesh woven from 0.28mm (0.011in) dia wire
Thermo Scientific Chemicals

Titanium gauze, 18 mesh woven from 0.28mm (0.011in) dia wire

CAS: 7440-32-6 | Ti | 47.87 g/mol
数量:
125 x 125 mm
250 x 250 mm
製品番号(カタログ番号) 043794.GN
または、製品番号043794-GN
価格(JPY)
-
数量:
125 x 125 mm
一括またはカスタム形式をリクエストする
仕様
外見Silver Gray guaze
CAS7440-32-6
ChEBICHEBI:33341
化学物質名または材質Titanium
Silver to Gray
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only