Thermo Scientific Chemicals

Diethyl phosphite, 96%, Thermo Scientific Chemicals

Catalog number: 043801.A1
1 kg, Each
Thermo Scientific Chemicals

Diethyl phosphite, 96%, Thermo Scientific Chemicals

Catalog number: 043801.A1
1 kg, Each
Quantity

Chemical Identifiers

CAS
762-04-9
IUPAC Name
diethoxy(oxo)-λ⁵-phosphanylium
Molecular Formula
C4H10O3P
InChI Key
LXCYSACZTOKNNS-UHFFFAOYSA-N
SMILES
CCO[P+](=O)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥ 95.0 %
Form
Liquid

Description

Diethyl phosphite used as intermediate for extraction agent and phosphoric acid esters. It is also used as solvent and acid acceptor in the synthesis of polypeptide. It is also used in the synthetic reaction. It is a precursor of phosphorous which is used for the synthesis of various organophosphorus compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text