3,3'-Diethylthiadicarbocyanine iodide
3,3'-Diethylthiadicarbocyanine iodide
3,3'-Diethylthiadicarbocyanine iodide
Thermo Scientific Chemicals

3,3'-Diethylthiadicarbocyanine iodide

As a solvent soluble laser dye | CAS: 514-73-8 | C23H24IN2S2+ | 519.483 g/mol
数量:
0.5 g
2 g
10 g
製品番号(カタログ番号) 043851.02
または、製品番号043851-02
価格(JPY)
-
数量:
0.5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS514-73-8
IUPAC Name3-ethyl-2-[(3E)-5-[(2E)-3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular FormulaC23H23IN2S2
InChI KeyMNQDKWZEUULFPX-UHFFFAOYSA-M
SMILES[I-].CCN1C(SC2=CC=CC=C12)=C/C=C/C=CC1=[N+](CC)C2=CC=CC=C2S1
さらに表示
lambda-max (UV-Vis)650 to 660 nm
Appearance (Color)Green
Formpowder`
Assay (unspecified)absorptivity at lamda-max: > 175,000 L /mol.cm
It is employed as a solvent soluble laser dye. DTDCi is used as fluorescent contrast agent in three-dimensional fluorescence lifetime tomography and as a dye for rapid genetic screening. It is a potential sensitive probe useful for fluorometric analysis of transferable membrane pores and effects of membrane potential on Na cotransports in eel intestinal brush-border membrane vesicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only