Thermo Scientific Chemicals

Sulfobromophthalein sodium salt hydrate

Catalog number: 043878.03
1 g, Each
Thermo Scientific Chemicals

Sulfobromophthalein sodium salt hydrate

Catalog number: 043878.03
1 g, Each
Quantity
Catalog number: 043878.03
also known as 043878-03
Price (USD)
Price: 32.40
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
71-67-0
IUPAC Name
disodium 2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzene-1-sulfonate
Molecular Formula
C20H8Br4Na2O10S2
InChI Key
GHAFORRTMVIXHS-UHFFFAOYSA-L
SMILES
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O
Appearance (Color)
White to off-white to grey to light brown
lambda-max (UV-Vis)
575-581 nm
Form
Powder
Loss on Drying
<7%

Description

Sulfobromophthalein sodium salt hydrate is a reagent for the quantitative determination of proteins, a dye for liver diagnostics in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text