Bismuth(III) isopropoxide, Thermo Scientific Chemicals
Bismuth(III) isopropoxide, Thermo Scientific Chemicals
Bismuth(III) isopropoxide, Thermo Scientific Chemicals
Bismuth(III) isopropoxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) isopropoxide, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
043938.06
または、製品番号043938-06
5 g
製品番号(カタログ番号) 043938.06
または、製品番号043938-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15049-67-9
IUPAC Namebismuth(3+) tris(propan-2-olate)
Molecular FormulaC9H21BiO3
InChI KeyKNPRLIQQQKEOJN-UHFFFAOYSA-N
SMILES[Bi+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
さらに表示
Appearance (Color)Grey to dark grey or dark grey-brown
Assay from Suppliers CofA≥95.0% (Gravimetirc)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only