Poly(vinylidene fluoride) - FAQs
View additional product information for Poly(vinylidene fluoride) - FAQs (044080.A3, 044080.36, 044080.A9)
1 product FAQs found
What is the average molecular weight of Poly(vinylidene fluoride)?
The average molecular weight of Poly(vinylidene fluoride) ranges from 250 and 450 kD.