Zirconium foil, 0.711mm (0.028in) thick, annealed, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium foil, 0.711mm (0.028in) thick, annealed, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
100 x 500 mm
Catalog number 044253.GH
Price (USD)/ Each
-
Quantity:
100 x 100 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormFoil, 0.711mm (0.028in) thick, annealed.
Assay from Supplier's CofA≥99.5% (metals basis excluding Hf)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only