Search
View additional product information for Silica gel desiccant, indicating, -6+16 mesh granules - FAQs (044389.A1, 044389.30, 044389.A7)
2 product FAQs found
Silica gel desiccant, indicating, -6+16 mesh granules gradually change from a deep blue in the activated condition to a pale pink in the saturated condition.
Yes, Silica gel desiccant, indicating, -6+16 mesh granules can be regenerated by heating at 121-232 degrees C until it returns to a blue color.