Thermo Scientific Chemicals

Barium oxide, 99.5% (metals basis excluding Sr), Sr <500ppm, Thermo Scientific Chemicals

Catalog number: 044440.22
100 g, Each
Thermo Scientific Chemicals

Barium oxide, 99.5% (metals basis excluding Sr), Sr <500ppm, Thermo Scientific Chemicals

Catalog number: 044440.22
100 g, Each
Quantity
Catalog number: 044440.22
also known as 044440-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1304-28-5
IUPAC Name
oxobarium
Molecular Formula
BaO
InChI Key
QVQLCTNNEUAWMS-UHFFFAOYSA-N
SMILES
O=[Ba]
Impurity content
Sr<500ppm
Total Metal Impurities
0.5% max.

Description

Barium oxide is used in cathode ray tubes, crown glass, and catalysts. It is used for the adsorption NO and NO2. It also plays an important role for NO2 storage and reduction. It acts as an efficient ethoxylation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text