Nickel etchant for electrodeposited nickel films - FAQs
View additional product information for Nickel etchant for electrodeposited nickel films, Thermo Scientific Chemicals - FAQs (044580.R0)
5 product FAQs found
What is the etch rate of Nickel etchant for electrodeposited nickel films at 40 degrees C?
Nickel etchant for electrodeposited nickel films has an etch rate of 50 Å/second at 40 degrees C.
How do I reduce undercutting of an etchant?
Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.
Do I need to dilute an etchant?
No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.
How do I reduce the etch rate of an etchant?
The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.
How do I increase the etch rate of an etchant?
The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.