Thermo Scientific Chemicals
HSG, Heat sink grease, Thermo Scientific Chemicals
Catalog number: 044589.5X
4 oz., Each
Thermo Scientific Chemicals
HSG, Heat sink grease, Thermo Scientific Chemicals
Catalog number: 044589.5X
4 oz., Each
Quantity
Catalog number: 044589.5X
also known as 044589-5X
Price (USD)
Price: 62.50
Online price: 53.65
Your price:
Quantity
-
Transition Temperature
1.75x10-3gramcal./cm2/sec./C/cm
Comment
Operating temperature range -60? to 200?C
Description
Thermally conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent, to reduce thermal contact resistance. Ideal for sputtering target applications.