Thermo Scientific Chemicals

Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals

Catalog number: 044646.22
100 g, Each
Thermo Scientific Chemicals

Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals

Catalog number: 044646.22
100 g, Each
Quantity
Catalog number: 044646.22
also known as 044646-22
Price (USD)
Price: 501.00
Online price: 425.65
Your price:
Quantity
-

Chemical Identifiers

CAS
409-21-2
IUPAC Name
methanidylidynesilylium
Molecular Formula
CSi
InChI Key
HBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES
[C-]#[Si+]
Impurity content
0.15% free Si max

Description

Silicon carbide can be used form very hard ceramics that are widely used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide as light-emitting diodes (LEDs) and detectors in early radios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text