Tetramethylammonium hydroxide, 2.38% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)
Tetramethylammonium hydroxide, 2.38% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)
Tetramethylammonium hydroxide, 2.38% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)
Thermo Scientific Chemicals

Tetramethylammonium hydroxide, 2.38% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)

CAS: 75-59-2 | C4H13NO | 91.154 g/mol
数量:
250 mL
1 L
1 gal.
4 x 1 gal.
製品番号(カタログ番号) 044940.LV
または、製品番号044940-LV
価格(JPY)
-
数量:
1 gal.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-59-2
IUPAC Nametetramethylazanium hydroxide
Molecular FormulaC4H13NO
InChI KeyWGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C
さらに表示
Total Metal Impurities0.0001% max.
Tetramethylammonium hydroxide is used as anisotropic etching of silicon, as a basic solvent in the development of acidic photo resist in the photolithography process, and is highly effective in stripping photo resist, and is used as a surfactant in the synthesis of ferrofluid, to inhibit nanoparticle aggregation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only