Tripotassium citrate monohydrate, 97%
Tripotassium citrate monohydrate, 97%
Tripotassium citrate monohydrate, 97%
Thermo Scientific Chemicals

Tripotassium citrate monohydrate, 97%

CAS: 6100-05-6 | C6H7K3O8 | 324.41 g/mol
製品番号(カタログ番号) 045038.0E
または、製品番号045038-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6100-05-6
IUPAC Nametripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Molecular FormulaC6H7K3O8
InChI KeyPJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILESO.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
さらに表示
FormGranular or chunks
Assay from Supplier's CofA≥96.0%
It is used in the laboratory as a buffering agent and chelator. In renal function studies, potassium citrate is used as an alkanizing agent where the citric acid is a metabolic substrate in renal cells. Studies have shown that potassium citrate may increase bone density, it is therefore used in research experiments related to the treatment of osteoporosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only