Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
Thermo Scientific Chemicals

Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol

CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol
数量:
5 mL
100 mL
500 mL
製品番号(カタログ番号) 045480.AP
または、製品番号045480-AP
価格(JPY)
-
数量:
500 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1184-61-8
IUPAC Nametetrakis(propan-2-ol) tin
Molecular FormulaC12H32O4Sn
InChI KeyVUDAJANAMZGVOP-UHFFFAOYSA-N
SMILES[Sn].CC(C)O.CC(C)O.CC(C)O.CC(C)O
さらに表示
Appearance (Color)Yellow to red
FormLiquid
Concentration (w/v)9.5-10.5%
Total Metal Impurities≤1%
Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only