Sulfuric acid, 15% fuming, 12-17% free SO{3}
Sulfuric acid, 15% fuming, 12-17% free SO{3}
Sulfuric acid, 15% fuming, 12-17% free SO{3}
Thermo Scientific Chemicals

Sulfuric acid, 15% fuming, 12-17% free SO{3}

CAS: 8014-95-7 | H2O7S2 | 178.13 g/mol
数量:
500 g
4 x 500 g
製品番号(カタログ番号) 045542.36
または、製品番号045542-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
仕様
定量パーセント範囲15% fuming, 12-17% free SO3
CAS8014-95-7
化学物質名または材質Sulfuric acid; 15% fuming
DOT情報Hazard Class: 8; Packaging Group: II
密度1.9
さらに表示
Sulfuric acid is a strong dehydrating agent and is applied in many applications like domestic acidic drain cleaners, electrolytes in lead-acid batteries and various cleansing agents. It is useful for mineral processing, fertilizer manufacture, petroleum purification, wastewater processing and chemical synthesis. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows a useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only