Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)
Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)
Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)
Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)
Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)
Thermo Scientific Chemicals

Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
数量:
100 g
500 g
2 kg
製品番号(カタログ番号) 045544.22
または、製品番号045544-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
さらに表示
Appearance (Color)Red-brown
FormPowder
Total Metal Impurities≤0.1%
Particle Size+170 mesh (≥90µm): ≤10% by weight
Particle Size-270 mesh (≤53µm): ≤10% by weight
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only