Thermo Scientific™

Praseodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific™

25 g, Each
Thermo Scientific™

Praseodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific™

25 g, Each
Quantity
Catalog number: 045910.14
also known as 045910-14
Price (USD)
Price: 739.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
7440-10-0
IUPAC Name
praseodymium
Molecular Formula
Pr
InChI Key
PUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES
[Pr]
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Packaged under Argon
RUO – Research Use Only

Figures