Aluminum oxide, basic, HPLC Flash Grade, Thermo Scientific Chemicals
Aluminum oxide, basic, HPLC Flash Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, basic, HPLC Flash Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 046071.22
also known as 046071-22
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
57.10 
Save 8.45 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
57.10 
Save 8.45 (15%)
Add to cart
Aluminum oxide, basic, HPLC Flash Grade, Thermo Scientific Chemicals
Catalog number046071.22
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
57.10 
Save 8.45 (15%)
-
Add to cart
Specifications
Boiling Point2980°C
Chemical Name or MaterialAluminum oxide
Melting Point2045°C
Name NoteBasic
CAS1344-28-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only