Thermo Scientific Chemicals

Sepabeads|r SP710, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 90 angstroms, Thermo Scientific Chemicals

Catalog number: 046555.A1
1 kg, Each
Thermo Scientific Chemicals

Sepabeads|r SP710, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 90 angstroms, Thermo Scientific Chemicals

Catalog number: 046555.A1
1 kg, Each
Quantity
Catalog number: 046555.A1
also known as 046555-A1
Price (USD)
Price: 224.00
Online price: 199.65
Your price:
Quantity
-
Assay from Supplier's CofA
Water Content 60-70%
Surface Area
ca. 900m²/g
Particle Size
250?m
Form
Beads

Description

Synthetic adsorbents for juice debittering and decolorizing. Also highest naringin capacity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Synthetic adsorbents for juice debittering and decolorizing
  • Also highest naringin capacity
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text