Thermo Scientific Chemicals

Indiumdraht, 1.0 mm (0.04 Zoll) Durchmesser, geglüht, Puratronic™, ≥99.998 % (Metallbasis), Thermo Scientific Chemicals

Katalognummer: 046661.G5
5 m, Each
Thermo Scientific Chemicals

Indiumdraht, 1.0 mm (0.04 Zoll) Durchmesser, geglüht, Puratronic™, ≥99.998 % (Metallbasis), Thermo Scientific Chemicals

Katalognummer: 046661.G5
5 m, Each
Menge
Katalognummer: 046661.G5
auch als 046661-G5 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
7440-74-6
IUPAC Name
indium
Molecular Formula
In
InChI Key
APFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES
[In]
Form
Wire, 1.0mm (0.04in) diameter, annealed.
Appearance (Color)
Silvery
Assay from Supplier's CofA
>99.998% (metals basis)

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text