Thermo Scientific Chemicals

Silver bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046784.06
5 g, Each
Thermo Scientific Chemicals

Silver bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog number: 046784.06
5 g, Each
Quantity
Catalog number: 046784.06
also known as 046784-06
Price (USD)
Price: 727.00
Online price: 617.65
Your price:
Quantity
-

Chemical Identifiers

CAS
189114-61-2
IUPAC Name
silver(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular Formula
C2AgF6NO4S2
InChI Key
HSYLTRBDKXZSGS-UHFFFAOYSA-N
SMILES
[Ag+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Form
Powder
Appearance (Color)
White to orange

Description

Silver bis(trifluoromethylsulfonyl)imide is used in the extraction of C5 - C8 linear alfa olefins using its silver salt to form room temperature ionic liquids. Further, it is utilized for the separation of olefins from paraffins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text