Thermo Scientific Chemicals

Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals

Catalog number: 046801.MD
250 mg, Each
Thermo Scientific Chemicals

Polyethylene glycol diacrylate, M.W. 8,000, Thermo Scientific Chemicals

Catalog number: 046801.MD
250 mg, Each
Quantity
Catalog number: 046801.MD
also known as 046801-MD
Price (USD)
Quantity
-

Chemical Identifiers

CAS
26570-48-9
IUPAC Name
ethane-1,2-diol; prop-2-enoic acid
Molecular Formula
C5H10O4
InChI Key
FCDZZFLRLVQGEH-UHFFFAOYSA-N
SMILES
OCCO.OC(=O)C=C
Appearance (Color)
Pale yellow to pale cream
Form
Granular solid
Assay from Supplier's CofA
≥95.0%
Comment
Inhibitor: Report type and quantity
Molecular Weight
8,000 (approx.)

Description

Polyethylene glycol diacrylate is used to prepare photopolymerized hydrogels which are used as scaffolds for tissue regeneration and other biological applications. It acts as an important raw material for the synthesis of poly(ethylene glycol) urethane-dimethacrylates (PEGUDM) by reacting with 2-isocyanatoethyl methacrylate. It contributes to chemical resistance, flexibility, adhesion, low shrinkage, abrasion resistance and impact strength.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text