n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles
n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles
n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles

CAS: 142-82-5 | C7H16 | 100.21 g/mol
製品番号(カタログ番号) 047191.AK
または、製品番号047191-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
さらに表示
Appearance (Color)Colorless
FormLiquid
Assay from Supplier's CofA≥98.5%
Residue on Evaporation≤5 ppm
Water Content≤10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only