Zirconium hydride, 99%, Thermo Scientific Chemicals
Zirconium hydride, 99%, Thermo Scientific Chemicals
Zirconium hydride, 99%, Thermo Scientific Chemicals
Zirconium hydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium hydride, 99%, Thermo Scientific Chemicals

数量:
10 g
50 g
250 g
製品番号(カタログ番号) 047203.30
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7704-99-6
IUPAC Nameλ²-zirconium(2+) dihydride
Molecular FormulaH2Zr
InChI KeyIUJYJRCMJZIAJY-UHFFFAOYSA-N
SMILES[H-].[H-].[Zr++]
さらに表示
Particle Size4.5 - 6.5 micron APS
FormPowder, -325 mesh
Assay (unspecified)≥98.5%
Particle Size4.5 - 6.5 micron APS
Assay (unspecified)≥98.5%
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only