Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Thermo Scientific Chemicals

Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)

For iodometric titrations | CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
数量:
500 mL
1 L
4 L
20 L
製品番号(カタログ番号) 047209.K7
価格(JPY)
-
数量:
4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
さらに表示
Appearance (Color)Colorless
FormLiquid
Concentration1.0% w/v ± 0.05% w/v
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only