Calcium carbonate, Puratronic™, 99.999% (metals basis)
Calcium carbonate, Puratronic™, 99.999% (metals basis)
Calcium carbonate, Puratronic™, 99.999% (metals basis)
Calcium carbonate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.999% (metals basis)

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 047235.14
または、製品番号047235-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
さらに表示
FormPowder
Total Metal Impurities10 ppm, max
Applied as neutralizing agent, filler, and extender in rubber, plastics, paints, opacifying agent in paper, putty, antacid, Portland cement, sulfur dioxide, removal from stack gases, allurgical flux, analytical chemistry, carbon dioxide generation, and determining halogens in organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only